missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters

Product Code. 10205774
Change view
Click to view available options
Model:
S9 DO Meter
Seven2Go Pro S8
Seven2Go S2
Seven2Go S3
Seven2Go S4
SevenGo Duo SG23
SevenGo Duo SG68
SevenGo Duo SG78
SevenGo Duo SG98
pH, ORP
Channels:
1
2
Product Type:
Conductivity Meter
DO Meter
DO Portable Meter
Meter with BOD Kit
Meter with Bioethanol Kit
Meter with Biotech Kit
Meter with Electrode Kit
Meter with Field Kit
Meter with Field Kit, 2m Cable
Meter with Field Kit, 5m Cable
Meter with Fluoride Kit
Meter with Food Kit
Meter with Light Kit
Meter with Standard Kit
pH/Conductivity Meter
pH/Ion/Cond Meter
pH/Ion/DO Meter
pH/Ion/DO/ORP Meter
pH/mV Meter
pH/mV and Ion Meter
Unit Size:
1 set
Each
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Channels Product Type unitSize
10205774 SevenGo Duo SG23 2 Meter with Electrode Kit 1 set
10379304 SevenGo Duo SG23 2 pH/Conductivity Meter Each
11967981 SevenGo Duo SG98 2 Meter with Field Kit, 2m Cable 1 set
11918011 SevenGo Duo SG98 2 Meter with Field Kit, 5m Cable 1 set
10422005 SevenGo Duo SG68 2 pH/Ion/DO Meter Each
10717315 SevenGo Duo SG68 2 Meter with Electrode Kit Each
10042274 SevenGo Duo SG78 2 Meter with Electrode Kit Each
10297814 SevenGo Duo SG78 2 Meter with Field Kit Each
15345073 Seven2Go S3 1 Conductivity Meter Each
15355073 Seven2Go S3 1 Meter with Standard Kit Each
15365073 Seven2Go S3 1 Meter with Field Kit Each
15375073 Seven2Go S3 1 Meter with Bioethanol Kit Each
15385073 Seven2Go S4 1 DO Meter Each
15395073 Seven2Go S4 1 Meter with Standard Kit Each
15305083 Seven2Go S4 1 Meter with Field Kit Each
15315093 S9 DO Meter 1 Meter with Standard Kit Each
15325093 S9 DO Meter 1 Meter with Field Kit Each
15335093 S9 DO Meter 1 Meter with BOD Kit Each
15395063 Seven2Go S2 1 pH/mV Meter Each
15305073 Seven2Go S2 1 Meter with Standard Kit Each
15315073 Seven2Go S2 1 Meter with Field Kit Each
15325073 Seven2Go S2 1 Meter with Food Kit Each
15335073 pH, ORP 1 Meter with Light Kit Each
15355083 Seven2Go Pro S8 1 pH/mV and Ion Meter Each
15375083 Seven2Go Pro S8 1 Meter with Field Kit Each
15385083 Seven2Go Pro S8 1 Meter with Biotech Kit Each
15395083 Seven2Go Pro S8 1 Meter with Fluoride Kit Each
15305093 S9 DO Meter 1 DO Portable Meter Each
10430665 SevenGo Duo SG78 2 pH/Ion/Cond Meter Each
11957981 SevenGo Duo SG98 2 pH/Ion/DO/ORP Meter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
This item is not returnable. View return policy
Product Code. 10205774 Supplier METTLER TOLEDO™ Supplier No. 51302601
Wastewater Testing Savings
Use promo code 28023
View all promotions
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.

  • Dust proof
  • Water proof
  • IP67 Rated
  • pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
  • Compatible with a wide range of IP67-rated electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Data storage available; capacity dependent on model
  • Export data to a printer or Excel file for Seven2Go Pro only
  • EasyDirect software available for data transfer (model dependent) for Seven2Go Pro only
  • Rubber holster available for single-handed measurements
  • uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Conductivity) ±0.5%
Accuracy (mV) ±1mV
Accuracy (pH) ±0.01
Accuracy (TDS) ±0.5%
Battery Type 4 x AA
Calibration Points 3 Points, 4 pre defined buffer group
Certifications/Compliance ISO 9001:2000
Data Management Details 99 GLP data records
Channels 2
Display Type LCD
IP Rating IP67
Measurement Modes ORP
Range (Conductivity) 0.01 μS/cm to 500 mS/cm
Range (mV) -1999 to 1999 mV
Range (pH) 0 to 14
Range (Resistivity) 0 to 100 Mohm/cm
Range (Salinity) 0 to 80 ppt
Range (TDS) 0.01 mg/L to 300 g/L
Resolution (pH) 0.01
Temperature Range (Metric) -5°C to +105°C
For Use With (Application) Determination of pH, mV, conductivity, TDS (total dissolved solids), salinity and specific resistance
Includes InLab Expert Pro ISM pH/temperature electrode; InLab 738-ISM conductivity/temperature electrode
Inputs pH: BNC/RCA, conductor: LTW 7-pin
Model SevenGo Duo SG23
Probe Type NTC
Product Type Meter with Electrode Kit
Resolution (Conductivity) 0.01/1
Resolution (mV) 1mV
Resolution (TDS) 0.01/1
Temperature Accuracy ±0.5°C
Temperature (English) Operating 30°F to 104°F
Temperature (Metric) Operating 0°C to 40°C
Temperature Resolution 0.1°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.