Learn More
METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters
Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.
575.00 € - 3460.00 €
Product Code | Brand | Product Type | Includes | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Includes | Model | Price | Quantity & Availability | |||
10205774
|
METTLER TOLEDO™
51302601 |
Electrode Kit | InLab Expert Pro ISM pH/temperature electrode; InLab 738-ISM conductivity/temperature electrode | Dual Channel; pH/ORP, conductivity |
1705.00 €
1 set |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10379304
|
METTLER TOLEDO™
51302600 |
Meter only (SG23) | Meter Only | Dual Channel; pH/ORP, conductivity |
1080.00 €
Each |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11957981
|
METTLER TOLEDO™
51302661 |
Meter Only | Meter Only | Dual Channel; pH/ORP/Ion, DO |
2015.00 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||
11967981
|
METTLER TOLEDO™
51302663 |
Meter Field Kit with Electrodes and Case | InLab OptiOx Dissolved oxygen/temperature electrode; InLab Expert Pro-ISM pH/temperature electrode | Dual Channel; pH/ORP/Ion, DO |
3345.00 €
1 set |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11918011
|
METTLER TOLEDO™
51302664 |
Meter Field Kit with Electrodes and Case | InLab OptiOx-5m Dissolved oxygen/temperature electrode; InLab Expert Go-5m-ISM pH/temperature electrode | Dual Channel; pH/ORP/Ion, DO |
3460.00 €
1 set |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10422005
|
METTLER TOLEDO™
51302610 |
Meter only | Meter Only | Dual Channel; pH/ORP/ion, DO |
1990.00 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||
10717315
|
METTLER TOLEDO™
51302611 |
Meter and Electrode kit, Includes 2 m cable and 2-electrode clip | InLab 605-ISM Dissolved Oxygen/ temperature electrode; InLab Expert Go-ISM pH / temperature electrode | Dual Channel; pH/ORP/Ion, DO |
2860.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10430665
|
METTLER TOLEDO™
51302620 |
Portable Meter | Meter Only | Dual Channel; pH/ORP/ion, conductivity |
1685.00 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||
10042274
|
METTLER TOLEDO™
51302621 |
Electrode Kit | InLab 738-ISM conductivity/temperature electrode; InLab Expert Go-ISM pH/temperature electrode | Dual Channel; pH/ORP/ion, conductivity |
2325.00 €
Each |
N/A | |||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10297814
|
METTLER TOLEDO™
51302622 |
Field Kit | InLab 738-ISM conductivity/temperature electrode; InLab Expert Go-ISM pH/temperature electrode | Dual Channel; pH/ORP/ion, conductivity |
2730.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15345073
|
METTLER TOLEDO™
30207954 |
Seven2Go S3 Conductivity Meter | Meter Only | Single channel; conductivity, TDS, salinity |
615.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15355073
|
METTLER TOLEDO™
30207955 |
Seven2Go S3 Conductivity Meter: Standard Kit | InLab 738-ISM conductivity/temperature electrode | Single channel; conductivity, TDS, salinity |
905.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15365073
|
METTLER TOLEDO™
30207956 |
Seven2Go S3 Conductivity Meter: Field Kit | InLab 738-ISM conductivity/temperature electrode; uGo carrying/protective case | Single channel; conductivity, TDS, salinity |
955.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15375073
|
METTLER TOLEDO™
30207957 |
Seven2Go S3 Conductivity Meter: Bioethanol Kit | InLab 725 conductivity/temperature electrode; uGo carrying/protective case | Single channel; conductivity, TDS, salinity |
1025.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15315083
|
METTLER TOLEDO™
30207961 |
S7 Conductivity Meter | Meter Only | Single channel; conductivity, TDS, salinity |
980.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15325083
|
METTLER TOLEDO™
30207962 |
S7 Conductivity Meter: Standard Kit | InLab 738-ISM conductivity/temperature electrode | Single channel; conductivity, TDS, salinity |
1300.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15335083
|
METTLER TOLEDO™
30207963 |
S7 Conductivity Meter: Field Kit | InLab 738-ISM conductivity/temperature electrode; uGo carrying/protective case | Single channel; conductivity, TDS, salinity |
1355.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15345083
|
METTLER TOLEDO™
30207873 |
S7 Conductivity Meter: USP/EP Kit | InLab 742-ISM conductivity/temperature electrode; uGo carrying/protective case | Single channel; conductivity, TDS, salinity |
1430.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15385073
|
METTLER TOLEDO™
30207958 |
Seven2Go S4 DO Meter | Meter only | Single channel; conductivity, TDS, salinity |
650.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15395073
|
METTLER TOLEDO™
30207959 |
Seven2Go S4 DO Meter: Standard Kit | InLab 605-ISM polarographic dissolved oxygen/ temperature electrode | Single channel; dissolved oxygen (DO) |
1100.00 €
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15305083
|
METTLER TOLEDO™
30207960 |
Seven2Go S4 DO Meter: Field Kit | InLab 605-ISM polarographic dissolved oxygen/ temperature electrode; uGo carrying/protective case | Single channel; dissolved oxygen (DO) |
1165.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15315093
|
METTLER TOLEDO™
30207971 |
S9 DO Meter: Standard kit | InLab OptiOx optical dissolved oxygen/ temperature electrode | Single channel; dissolved oxygen (DO) |
1755.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15325093
|
METTLER TOLEDO™
30207972 |
S9 DO Meter: Field kit | InLab OptiOx optical dissolved oxygen/ temperature electrode; uGo carrying/protective case | Single channel; dissolved oxygen (DO) |
1810.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15335093
|
METTLER TOLEDO™
30207939 |
S9 DO Meter: BOD kit | InLab OptiOx optical dissolved oxygen/ temperature electrode; uGo carrying/protective case; BOD adapter | Single channel; dissolved oxygen (DO) |
1870.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15395063
|
METTLER TOLEDO™
30207949 |
Seven2Go S2 pH/mV Meter | Meter Only | Single channel; pH, ORP |
575.00 €
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15305073
|
METTLER TOLEDO™
30207950 |
Seven2Go S2 pH/mV Meter: Standard Kit | InLab Expert Go-ISM pH/temperature electrode | Single channel; pH, ORP |
715.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15315073
|
METTLER TOLEDO™
30207951 |
Seven2Go S2 pH/mV Meter: Field Kit w/ pH probe | InLab Expert Go-ISM pH/temperature electrode; uGo carrying/protective case | Single channel; pH, ORP |
775.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15325073
|
METTLER TOLEDO™
30207952 |
Seven2Go S2 pH/mV Meter: Food Kit | InLab Solids Go-ISM pH/temperature electrode | Single channel; pH, ORP |
1005.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15335073
|
METTLER TOLEDO™
30207953 |
Seven2Go S2 pH/mV Meter: Light Kit | inLab Versatile Pro pH/temperature electrode | Single channel; pH, ORP |
630.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15355083
|
METTLER TOLEDO™
30207874 |
Seven2Go Pro S8 pH/mV and Ion Meter | Meter Only | Single channel; pH, Ion, ORP |
920.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15375083
|
METTLER TOLEDO™
30207877 |
Seven2Go Pro S8 pH/mV and Ion Meter: Field Kit | InLab Expert Go-ISM pH/temperature electrode; uGo carrying/protective case | Single channel; pH, Ion, ORP |
1160.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15385083
|
METTLER TOLEDO™
30207878 |
Seven2Go Pro S8 pH/mV and Ion Meter: Biotech Kit | InLab Routine Pro-ISM pH/temperature electrode | Single channel; pH, Ion, ORP |
1255.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15395083
|
METTLER TOLEDO™
30207879 |
Seven2Go Pro S8 pH/mV and Ion Meter: Fluoride Kit | perfectION Fluoride combination Fluoride ion selective electrode | Single channel; pH, Ion, ORP |
1575.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15305093
|
METTLER TOLEDO™
30207970 |
DO Portable S9-Meter | Meter Only | DO Portable Meter |
1025.00 €
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Dust proof
- Water proof
- IP67 Rated
- pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
- Compatible with a wide range of IP67-rated electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Data storage available; capacity dependent on model
- Export data to a printer or Excel file for Seven2Go Pro only
- EasyDirect software available for data transfer (model dependent) for Seven2Go Pro only
- Rubber holster available for single-handed measurements
- uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters
Specifications
ISO 9001, ISO 14001 | |
IP 67 |