Learn More
Mettler Toledo™ Temperature Calibration Kits
Product Code. 10588754 Shop All Mettler Toledo Products
Click to view available options
:
Without Certificate
Product Code. 10588754
Brand: Mettler Toledo™ 214455
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with halogen moisture analyzers. Mettler Toledo Temperature Calibration Kits are used to adjust halogen drying units to ensure consistent results.
  • Adjustment disc with precision thermometer
  • Sample pan handler
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Halogen Moisture Analyzer
Includes Adjustment disc with precision thermometer, sample
Type Temperature Calibration Kit