missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Mevalonolactone-d3 (methyl-d3), CDN
Click to view available options
Quantity:
0.025 g
Unit Size:
0.03g
Specifications
Specifications
| Chemical Name or Material | D,L-Mevalonic Acid Lactone-d3 |
| CAS | 61219-76-9 |
| Molecular Formula | C6 2H3 H7 O3 |
| Quantity | 0.025 g |
| InChI Formula | InChI=1 S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C1(O)CCOC(=O)C1 |
| IUPAC Name | 4-hydroxy-4-(trideuteriomethyl)oxan-2-one |
| Molecular Weight (g/mol) | 133.16 |
| Formula Weight | 133.0818 g/mol |
| Percent Purity | 99 atom % D, min 97% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction