missing translation for 'onlineSavingsMsg'
Learn More

Mevastatin, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18085762
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18085762 50 mg 50mg
18014042 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085762 Supplier Tocris Bioscience Supplier No. 1526/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

HMG-CoA reductase inhibitor

Inhibitor of HMG-CoA reductase; decreases cholesterol biosynthesis, in vitro and in vivo. Induces apoptosis, arrests cancer cells in G1 phase and downregulates cdk 2, 4, and 6, cyclin D1 and E1, p21 and p27. Inactive lactam prodrug of mevastatin hydroxy acid, naturally bioactivated in vivo following oral administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73573-88-3
Molecular Formula C23H34O5
Molecular Weight (g/mol) 390.52
MDL Number MFCD05662341
InChI Key AJLFOPYRIVGYMJ-INTXDZFKSA-N
Synonym mevastatin, compactin, mevastatinum, mevastatina, mevastatine, mevastatinum inn-latin, antibiotic ml 236b, mevastatin inn, compactin penicillium, mevastatine inn-french
PubChem CID 64715
ChEBI CHEBI:34848
IUPAC Name (1S,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1CCC=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Quantity 50 mg
Formula Weight 390.52
Percent Purity >98%
Chemical Name or Material Mevastatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.