missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences MEXi-TM

IBA Lifesciences GmbH MEXi-TM Transfection Medium is an efficient and cost effective mammalian protein expression system.

Brand:  IBA Lifesciences 2-6011-010

Additional Details : Weight : 1.00000kg

Product Code. 15533087

  • 118.00 € / 1000mL
Estimated Shipment: 19-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MEXi-TM transfection medium is a serum-free, chemically defined medium especially developed for transient transfection and growth of MEXi-293E cells.

It is part of the MEXi mammalian expression system. The MEXi system was developed to offer an affordable mammalian protein expression system coming along with all essential components:
• MEXi-293E cell line
• A variety of pDSG expression vectors
• MEXi-CM culture medium
• MEXi-TM transfection medium Further advantages:
• Transient expression for time- and cost-efficient protein production
• Multiple expression vectors with different features
• Optimized for Twin-Strep-tag™:Strep-Tactin™XT system for highly pure proteins
• IP-free system
Specifications

Specifications

1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers