missing translation for 'onlineSavingsMsg'
Learn More

MFG Tray Chemical-Resistant Multipurpose Trays and Pans

Product Code. 13074897 Shop All Cole-Parmer Products
Change view
Click to view available options
Dimensions (L x W x H):
18 x 14 x 1 in.
Type:
Multi-purpose Tray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Type Unit Size
13074897 18 x 14 x 1 in. Multi-purpose Tray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13074897 Supplier MFG Tray Supplier No. 0671582
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills

Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills

  • Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
  • Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
  • High load-bearing capacity
  • Excellent dimensional stability Easily steam cleaned

Order Info

Available in 1 in. deep (2.5cm) trays, shallow pans from 1 1/4 to 2 3/4 in. deep (3 to 7cm), and ventilated stacking pans from 2 3/4 to 4 1/8 in. deep (7 to 10.5cm).

Specifications

Dimensions (L x W x H) 18 x 14 x 1 in.
Color White
Height (Metric) 2.5 cm
Length (Metric) 46 cm
Material Fiberglass
Type Multi-purpose Tray
Width (Metric) 35.5 cm
For Use With (Application) Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Length (English) 18 in.
Height (English) 1 in.
Shape Rectangular
Temperature (English) -60°F to +250°F
Temperature (Metric) -51°C to +121°C
Width (English) 14 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.