missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MG 132

Product Code. 15465519
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465519 1 mg 1mg
15455519 5 mg 5mg
15475519 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15465519 Supplier Thermo Scientific Chemicals Supplier No. J63250.M^
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of calpain proteases | CAS: 133407-82-6475.63 g/mol

MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.

Solubility
Soluble in ethanol, chloroform, methanol, DMSO.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133407-82-6

Specifications

Color White
Content And Storage Store at -20°C
Format Crystalline Solid
Molecular Weight (g/mol) 475.63
Product Type MG 132
Quantity 1 mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.