missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ MG 132

Product Code. 18054562
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18054562 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18054562 Supplier R&D Systems™ Supplier No. 1748/5
  • ONLINE EXCLUSIVE PRICE
    233.27 €
    208.95 € / 5mg
    Save 24.32 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Proteasome and calpain inhibitor. Inhibits NF-κB activation

MG 132 is a potent cell-permeable inhibitor of proteasome (IC50 = 100 nM) and calpain (IC50 = 1.2 μM). MG 132 inhibits TNF-α-induced NF-κB activation and IκBα degradation. Induces neurite outgrowth in PC12 cells and has anticancer properties in vitro. MG 132 inhibits SARS-CoV-2 Mpro in vitro (IC50 = 3.9 μM) and inhibits viral replication. Also enhances adeno-associated viral transduction efficiency of human cell lines by around 50-fold.
TRUSTED_SUSTAINABILITY

Specifications

CAS 133407-82-6
Quantity 5 mg
Product Line Tocris
Product Type MG 132
Purity >95%
Molecular Weight (g/mol) 475.63
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.