Learn More
WTW™ MG 2.5 E Replacement Vessel
Product Code. 10064844
Change view
Click to view available options
Volume (Metric):
2500 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
10064844 2500 mL Each
1 options
Product Code. 10064844 Supplier WTW™ Supplier No. 209512
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 2500 mL