missing translation for 'onlineSavingsMsg'
Learn More

MG 624, Tocris Bioscience™

Product Code. 30001438
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001438 50 mg 50mg
18035292 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30001438 Supplier Tocris Bioscience Supplier No. 1356/50
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

α7 neuronal nicotinic receptor antagonist

A selective antagonist for neuronal α7 subunit- containing nAChR subtypes. Inhibits α-Bgtx binding to chick α7 and α4β2 subtypes with Ki values of 106 nM and 84 μM respectively.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77257-42-2
Molecular Formula C22H30INO
Molecular Weight (g/mol) 451.392
InChI Key RDTKUZXIHMTSJO-UEIGIMKUSA-M
Synonym stilonium iodide, stilonio ioduro dcit, stilonii iodidum inn-latin, iodure de stilonium inn-french, ioduro de estilonio inn-spanish, 2-p-styrylphenoxy ethyl triethylammonium iodide, triethyl 2-p-styrylphenoxy ethyl ammonium iodide, ioduro di trietil-beta-4-stilbenossi-etil ammonio italian, ammonium, 2-p-styrylphenoxy ethyl triethyl-, iodide, dsstox_cid_25781
PubChem CID 6433339
IUPAC Name triethyl-[2-[4-[(E)-2-phenylethenyl]phenoxy]ethyl]azanium;iodide
SMILES CC[N+](CC)(CC)CCOC1=CC=C(C=C1)C=CC2=CC=CC=C2.[I-]

Specifications

Quantity 50 mg
Formula Weight 451.39
Percent Purity >98%
Chemical Name or Material MG 624
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.