missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences™ MHC I-Strep H-2 Db; gp33 LCMV (KAVYNFATM); Fused to Twin-Strep-tag™

Product Code. 17540635 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 17540635

Brand: IBA Lifesciences™ 67016001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MHC Class I molecule fused to a streptavidin derivative and refolded with a peptide.

MHC I-Strep H-2 Db; gp33 LCMV (KAVYNFATM) is a recombinantly expressed MHC Class I molecule that presents a peptide from a lymphocytic choreomeningitis virus (LCMV) glycoprotein 33 (gp33). Our MHC I Streptamer™ approach is based on our Strep-tag™ technology and facilitates reversible antigen-specific T cell labeling. A Twin-Strep-tag™ fused to the MHC I-Strep permits multimerization of MHC I complexes on a Strep-Tactin™ backbone, resulting in the formation of MHC I Streptamers.

For multimerizing MHC I-Streps, Strep-Tactin™ TACS Agarose columns, Strep-Tactin™ Magnetic Microbeads or fluorophore-conjugated Strep-Tactin™ can be used. This allows affinity chromatographic or magnetic isolation or cell staining for flow cytometry analysis and sorting (FACS), depending on the experimental requirements.

  • Addition of biotin reverses multimerization of MHC Class I molecules, leading to dissociation from target cells
  • Resulting label-free cells are highly viable and fully functional

Specifications

Formulation Dissolved in DPBS
Workflow Step Cell Isolation
Quantity 10 μg
Target Antigen-specific T Cell
Species Human
Sufficient For 0.2 μg per 2 x 106 Total Cells
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.