Learn More
IBA Lifesciences™ MHC I-Strep HLA-A*0201; HER-2/neu (KIFGSLAFL); Fused to Twin-Strep-tag™
Shop All IBA Lifesciences ProductsDescription
MHC I-Strep HLA-A*0201; HER-2/neu (KIFGSLAFL) is a recombinantly expressed MHC Class I molecule that presents a peptide from a human epidermal growth factor receptor 2. Our MHC I Streptamer™ approach is based on our Strep-tag™ technology and facilitates reversible antigen-specific T cell labeling. A Twin-Strep-tag™ fused to the MHC I-Strep permits multimerization of MHC I complexes on a Strep-Tactin™ backbone, resulting in the formation of MHC I Streptamers.
For multimerizing MHC I-Streps, Strep-Tactin™ TACS Agarose columns, Strep-Tactin™ Magnetic Microbeads or fluorophore-conjugated Strep-Tactin™ can be used. This allows affinity chromatographic or magnetic isolation or cell staining for flow cytometry analysis and sorting (FACS), depending on the experimental requirements.
- Addition of biotin reverses multimerization of MHC Class I molecules, leading to dissociation from target cells
- Resulting label-free cells are highly viable and fully functional
Specifications
Specifications
| Formulation | Dissolved in DPBS |
| Workflow Step | Cell Isolation |
| Quantity | 10 μg |
| Target | Antigen-specific T Cell |
| Species | Human |
| Sufficient For | 0.2 μg per 2 x 106 Total Cells |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.