Learn More
IBA Lifesciences MHC I-Strep HLA-A*2402; CMV pp65 (QYDPVAALF)
MHC class I molecule fused to a Twin-Strep-tag™ and refolded with a peptide
Brand: IBA Lifesciences 6-7028-001
Additional Details : Weight : 0.00010kg
Description
Specifications
40 μL | |
Fluorescent reagent for staining, magnetic T cell labeling and isolation |