missing translation for 'onlineSavingsMsg'
Learn More

Mianserin hydrochloride

Product Code. 15424159
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424159 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15424159 Supplier Thermo Scientific Alfa Aesar Supplier No. J61570.MC
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21535-47-7 | C18H21ClN2 | 300.83 g/mol

Mianserin hydrochloride is used as an antidepressant, antagonist, inverse agonist at 5-HT2 serotonin receptors, also blocks the H1 histamine receptor and the α2 adrenoceptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mianserin hydrochloride is used as an antidepressant, antagonist, inverse agonist at 5-HT2 serotonin receptors, also blocks the H1 histamine receptor and the α2 adrenoceptor.

Solubility
Soluble in water, ethano
TRUSTED_SUSTAINABILITY

Specifications

CAS 21535-47-7
Color White
Molecular Formula C18H21ClN2
MDL Number MFCD00055072
Quantity 100 mg
Merck Index 14,6172
Synonym mianserin hydrochloride, mianserine hydrochloride, mianserin hcl, athymil, bolvidon, tolvon, tolvin, org gb 94, lerivon
Solubility Information Soluble in water,ethanol,methanol,and dimethylformamide.
InChI Key YNPFMWCWRVTGKJ-UHFFFAOYNA-N
SMILES [H+].[Cl-].CN1CCN2C(C1)C1=CC=CC=C1CC1=CC=CC=C21
IUPAC Name hydrogen 5-methyl-2,5-diazatetracyclo[13.4.0.0²,⁷.0⁸,¹³]nonadeca-1(19),8,10,12,15,17-hexaene chloride
Molecular Weight (g/mol) 300.83
PubChem CID 68551
ChEBI CHEBI:31843
Formula Weight 300.83
Physical Form Powder
Chemical Name or Material Mianserin hydrochloride
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.