missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Miconazole nitrate, 98+%, Thermo Scientific™

Inhibits cytochrome P450-dependent 14a-demethylase

Brand:  Thermo Scientific Alfa Aesar J60796.06

Quantity 5g

 View more versions of this product

Product Code. 15493499

  • ONLINE EXCLUSIVE PRICE
    131.00 €
    128.00 € / 5g
    Save 3.00 €
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

22832-87-7
479.14
MCCACAIVAXEFAL-UHFFFAOYNA-N
68553
O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1
C18H15Cl4N3O4
MFCD00058161
miconazole nitrate, albistat, andergin, aflorix, conofite, florid, micatin, gyno-monistat, epi-monistat, gyno-daktar
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole; nitric acid

Specifications

22832-87-7
MFCD00058161
14,6178
Slightly soluble in water; Soluble in propylene glycol or pyridine.
O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1
479.14
479.15
(+/-)-Miconazole nitrate
C18H15Cl4N3O4
5g
miconazole nitrate, albistat, andergin, aflorix, conofite, florid, micatin, gyno-monistat, epi-monistat, gyno-daktar
MCCACAIVAXEFAL-UHFFFAOYNA-N
1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole; nitric acid
68553
≥98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.