missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miconazole Nitrate, TRC
Specifications
Specifications
| Chemical Name or Material | Miconazole Nitrate |
| CAS | 22832-87-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H14 Cl4 N2 O . H N O3 |
| Quantity | 5 g |
| Synonym | 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-, mononitrate (9CI), Imidazole, 1-[2,4-dichloro-β-[(2,4-dichlorobenzyl)oxy]phenethyl]-, mononitrate (8CI), (±)-Miconazole nitrate, Aflorix, Albistat, Andergin, Antifungal Cream, Brentan, Conoderm, Conofite, Dactrine, Daktar, Daktarin, Daktarin talc, Deralbine, Dermonistat, Ecobi, Epi-Monistat, Florid, Fungiderm, Fungisdin, Gyno-Daktarin, Gyno-Monistat, Hi-Pick, Loptrimin AF Jock-Itch Powder Aerosol, Lotrimin AF Powder, Lotrimin AF Powder Aerosol, Lotrimin AF Spray Liquid, Micatin, Miconal, Miconaz, Miconazole nitrate, Micotef, Monistat, Monistat Cream and Suppositories, Monistat-Derm, NSC 169434, Priconazole, Prilagin, R 14889, Vodol, Zeasorb AF, Zimycan, [2,4-Dichloro-β-(2,4-dichlorobenzyloxy)phenethyl]imidazole nitrate |
| InChI Formula | InChI=1S/C18H14Cl4N2O.HNO3/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4) |
| SMILES | O[N+](=O)[O-].Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)c(Cl)c1 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid |
| Molecular Weight (g/mol) | 479.14 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?