missing translation for 'onlineSavingsMsg'
Learn More

Miconazole Nitrate, TRC

Product Code. 30469943
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30469943 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30469943

Brand: Toronto Research Chemicals TRCM342500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Miconazole Nitrate
CAS 22832-87-7
Purity Grade Notes HPLC
Molecular Formula C18 H14 Cl4 N2 O . H N O3
Quantity 100 mg
Synonym 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-, mononitrate (9CI), Imidazole, 1-[2,4-dichloro-β-[(2,4-dichlorobenzyl)oxy]phenethyl]-, mononitrate (8CI), (±)-Miconazole nitrate, Aflorix, Albistat, Andergin, Antifungal Cream, Brentan, Conoderm, Conofite, Dactrine, Daktar, Daktarin, Daktarin talc, Deralbine, Dermonistat, Ecobi, Epi-Monistat, Florid, Fungiderm, Fungisdin, Gyno-Daktarin, Gyno-Monistat, Hi-Pick, Loptrimin AF Jock-Itch Powder Aerosol, Lotrimin AF Powder, Lotrimin AF Powder Aerosol, Lotrimin AF Spray Liquid, Micatin, Miconal, Miconaz, Miconazole nitrate, Micotef, Monistat, Monistat Cream and Suppositories, Monistat-Derm, NSC 169434, Priconazole, Prilagin, R 14889, Vodol, Zeasorb AF, Zimycan, [2,4-Dichloro-β-(2,4-dichlorobenzyloxy)phenethyl]imidazole nitrate
InChI Formula InChI=1S/C18H14Cl4N2O.HNO3/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4)
SMILES O[N+](=O)[O-].Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)c(Cl)c1
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
Molecular Weight (g/mol) 479.14
Formula Weight 476.9817
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.