missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell MICRO-TOUCH™ Micro-Thin™ White Nitrile Gloves
Description
- ASTM D6978, ASTM D6319 certified
- Non-sterile glove made with a thinner formulation to provide high tactile sensitivity and dexterity
- High quality manufacturing ensures this glove will hold up for a variety of clinical procedures
- High tactile sensitivity is ideal for palpating veins and starting IVs
- Textured surface of the glove promotes sure handling of materials and instruments
- 200 gloves/box reduces restocking and reorders
- For use in chemotherapy and healthcare applications
Specifications
Specifications
| Certifications/Compliance | ASTM D6978, ASTM D6319, CE 0493, FDA21 CFR 177-2600-US Food Contact Approved, ISO 13485, ISO 9001 |
| Certifications/Compliance (Europe) | EN 16523-1, EN 374-2 and -4, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016, EN 420:2003 + A1:2009 |
| Accelerator Free | No |
| Applications | Healthcare |
| AQL | 1.5 |
| Color | White |
| Disposable | Single-use |
| Exterior Finish | Fingertip Textured |
| Finger Thickness (English) | 3.1 mil |
| Finger Thickness (Metric) | 0.08 mm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?