missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICRO-TOUCH™ Nitrile Gloves

Product Code. 17823802
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17823802 Medium Pack of 100
17863702 X-Small Pack of 100
17843802 Small Pack of 100
17803642 Large Pack of 100
17803752 X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 17823802 Supplier Ansell Supplier No. 313029080
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

MICRO-TOUCH™ Nitrile Gloves feature a thin, yet durable nitrile construction to provide healthcare professionals the reliable protection and tactility they need. These gloves are approved for chemotherapy use.

  • Designed with a thin, yet durable nitrile formulation for dependable protection
  • Royal blue color provides greater contrast than standard light blue nitrile gloves
  • Approved for use with chemotherapy drugs

Specifications

Applications Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 5.1 mil
Finger Thickness (Metric) 0.13 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Size Medium
Quantity 100 Gloves/Box, 10 Boxes/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.