missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell MICRO-TOUCH White Nitrile
Description
- Protects from latex Type 1 allergies
- 150-count packaging reduces waste
- Textured fingertips for secure handling of medical instruments
- Polymer inner coated for easy donning
- Tested for use with chemotherapy drugs
Specifications
Specifications
| Certifications/Compliance (Europe) | ASTM D6319, CE 0493, Category III, Medical Device Regulation (EU) 2017/745 Class I, EN 455 1-4, EN 421:2010, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, FDA21 CFR 177-2600-US Food Contact Approved, ISO 9001, ISO 10993-10:2002, ISO 13485, Food Type: Rubber, UKCA |
| Chemotherapeutic Rated | Yes, in accordance with ASTM D6978 (Not listed in the US FDA 510k) |
| Accelerator Free | Yes |
| Applications | Healthcare |
| AQL | 1.5 |
| Color | White |
| Disposable | Single-use |
| Exterior Finish | Fingertip Textured |
| Finger Thickness (English) | 3.9 mil |
| Finger Thickness (Metric) | 0.1 mm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?