Learn More
Cytiva Microarray Hybridization Solution v2.0
Product Code. 10762047 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Piece
Unit Size:
Each
Product Code. Quantity unitSize
10762047 1 Piece Each
1 options
Product Code. 10762047
Brand: Cytiva RPK0325
Uses a blend of proprietary components to provide quality automated and manual slide hybridizations. Cytiva Microarray Hybridization Solution v2.0 features hybridization rate enhancers that improve the detection of rare targets, plus blocking agents that minimize nonspecific background.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Piece