missing translation for 'onlineSavingsMsg'
Learn More

Microbiologics™ EZ-Accu Shot™ Microorganism Preparations

Product Code. 11936775
Encompass_Preferred
Change view
Click to view available options
Species:
Aspergillus brasiliensis
Bacillus subtilis
Clostridium sporogenes
Escherichia coli
Pseudomonas aeruginosa
Staphylococcus aureus
ATCC®:
11437™*
6538™*
8739™*
9027™*
ATCC 16404
ATCC 6633
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Species ATCC® unitSize
11936775 Aspergillus brasiliensis ATCC 16404 Each
11966775 Escherichia coli 8739™* Each
11986805 Pseudomonas aeruginosa 9027™* Each
11996805 Staphylococcus aureus 6538™* Each
11946775 Bacillus subtilis ATCC 6633 Each
11976805 Clostridium sporogenes 11437™* Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11936775 Supplier Microbiologics™ Supplier No. 0392A
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Product warranty and peel-off Certificate of Assay.

For easy growth promotion testing of culture media

Each lyophilized microorganism preparation is designed to deliver less than 100CFU per 0.1mL of hydrated suspension as recommended by the USP/JP/Ph. Eur. With little to no prep time required and up to eight hours of stability, EZ-Accu Shot adds efficiency and flexibility to QC lab schedules. Each 1.2mL vial of hydrated suspension offers 10 inocula. For added convenience, a peel-off Certificate of Assay is included for quality control documentation.

  • Instant dissolve, no pre-incubation for minimal prep time
  • One pellet per vial eliminates the need for forceps, reducing the risk of contamination
  • Eight-hour stability extends time and flexibility to perform tests
  • Precise CFU count ensures accurate results
  • Easy storage with no dry ice required, reducing shipping costs
  • Peel-off Certificate of Assay for complete, convenient documentation
  • Online Certificate of Analysis for documentation of phenotypic, macroscopic and microscopic test results
  • Traceable to USP reference culture or assured quality

Order Info

Each kit contains five vials of a single enumerated microorganism (one lyophilized pellet per vial) and five vials of hydrating fluid (1.2mL per vial).

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Specifications

ATCC® ATCC 16404
Form Pellets
Product Type EZ-Accu Shot
Species Aspergillus brasiliensis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.