Learn More
Ansell™ Microchem™ 3000 Coveralls with Hood and Integrated Socks
Product Code. 15566594
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 15
Pack of 20
Product Code. 15566594
Brand: Ansell™ YE30W0012205
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MICROCHEM™ 3000 Coveralls with Hood and Integrated Socks is one of the lightest and most comfortable chemical protective garments on the market today. This durable multi-layer fabric provides an extremely effective barrier against both inorganic chemicals and biological hazards.
  • Coverall with 2-piece hood
  • Integrated socks with boot overflap
  • Double zip closure
  • Double cuffs
  • Elasticated hood, waist, double cuffs
  • Excellent liquid penetration resistance and barrier to fine particulates (>0.01 microns)
  • Moisture vapour permeable (“breathable”) to help reduce the risk of heat stress
  • Silicone free for paint spraying applications
  • Low linting to reduces the risk of fibre contamination in some critical areas
  • Optimised body fit for wearer comfort and safety
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size X-Large
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Waist Style Elastic
Ankle Style Elastic, Attached Socks, Boot Overflaps
Cuff Style Double, Elastic
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper
Seam Type Welded
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN1149-5
Description Hood, integrated socks with boot overflap
No. of Pockets None
Style Includes Hood, Integrated socks with boot overflap