missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Microchem™ 3000 PAPR Encapsulated Coveralls

Product Code. 15546584
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15546584 Medium Pack of 4
15566584 X-Large Pack of 4
15556584 Large Pack of 4
15576584 2X-Large Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15546584 Supplier Ansell™ Supplier No. YE30T0070103

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides head and body protection from hazardous substances. MICROCHEM™ 3000 PAPR Encapsulated Coveralls feature a soft and flexible 3-layer fabric, strong ultrasonically welded seams and an effective chemical barrier against most inorganic chemicals.

  • Fully encapsulated coverall, for use in combination with Scott Proflow PAPR fan unit, breathing hose and filters.
  • Double elasticated cuffs, attached socks with boot over-flap.
  • Emergency rip cord attached to left chest shoulder and left back shoulder as worn
  • Multi-layer barrier fabric effective against numerous chemicals
  • Highly visible bright yellow for improved worker safety
  • Lightweight yet durable
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN 12941, EN1149-5
Color Yellow
Hood Type Fully Encapsulated
Material Microporous Polypropylene Laminate
Size Medium
Ankle Style Attached socks, Boot overflaps
Back Style Accommodates Scott Proflow 2SC/EX PAPR fan unit, breathing hoses, and filters
Collar Type Air permeable SMS
Cuff Style Double, Elastic
Description Emergency rip cord
For Use With (Equipment) Scott Proflow 2SC/EX PAPR fan unit, breathing hoses, and filters
No. of Pockets None
Product Type Encapsulated coveralls
Seam Type Stitched, Taped
Style Includes Emergency rip cord
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.