Learn More
Ansell™ Microchem™ 4000 Coveralls with Hood
Product Code. 15506444 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15506444 X-Large Pack of 10
15576434 Small Pack of 10
15586434 Medium Pack of 10
15596434 Large Pack of 10
15516444 2X-Large Pack of 10
15526444 3X-Large Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15506444 Supplier Ansell™ Supplier No. GR40T0011105
  • ONLINE EXCLUSIVE PRICE
    1036.40 €
    673.52 € / Pack of 10
    Save 362.88 €
    35% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides an exceptional barrier against organic and inorganic chemicals and biological agents. Ansell Microchem™ 4000 Coveralls with Hood is constructed of a unique multi-layer fabric known for its lightwieght, yet robust textile feel.
  • Coverall with elasticated 2-piece hood and front entry double-zip system.
  • Elasticated double cuffs (knitted inner cuff), waist and ankles.
  • Permeation tested against over 180 chemicals, including chemical warfare agents
  • Textile-like inner improves wearer acceptance
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Green
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Double, Elastic
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper Front Entry
Seam Type Taped, Welded
Resistance Chemicals
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN1149-5
No. of Pockets None
Style Includes Hood