Learn More
Ansell MICROCHEM™ 6000
Product Code. 17190915| Product Code. | Size | Style | unitSize |
|---|---|---|---|
| 17190915 | 3X-Large | O60T802E | Each |
| 17100915 | Large | O60T801B | Each |
| 17100925 | Small | O60T803E | Each |
| 17100945 | Large | O60T809E | Each |
| 17100955 | Small | O60A812E | Each |
| 17100965 | 2X-Large | O60A813E | Each |
| 17100975 | Large | O60A818E | Each |
| 17110915 | X-Large | O60T801B | Each |
| 17110925 | Medium | O60T803E | Each |
| 17110945 | X-Large | O60T809E | Each |
| 17110955 | Medium | O60A812E | Each |
| 17110965 | 3X-Large | O60A813E | Each |
| 17110975 | X-Large | O60A818E | Each |
| 17120915 | 2X-Large | O60T801B | Each |
| 17120925 | Large | O60T803E | Each |
| 17120935 | Small | O60T808E | Each |
| 17120945 | 2X-Large | O60T809E | Each |
| 17120955 | Large | O60A812E | Each |
| 17120975 | 2X-Large | O60A818E | Each |
| 17130915 | 3X-Large | O60T801B | Each |
| 17130925 | X-Large | O60T803E | Each |
| 17130935 | Medium | O60T808E | Each |
| 17130945 | 3X-Large | O60T809E | Each |
| 17130955 | X-Large | O60A812E | Each |
| 17130975 | 3X-Large | O60A818E | Each |
| 17140915 | Small | O60T802E | Each |
| 17140925 | 2X-Large | O60T803E | Each |
| 17140935 | Large | O60T808E | Each |
| 17140955 | 2X-Large | O60A812E | Each |
| 17140975 | Small | O60A819E | Each |
| 17150915 | Medium | O60T802E | Each |
| 17150925 | 3X-Large | O60T803E | Each |
| 17150935 | X-Large | O60T808E | Each |
| 17150955 | 3X-Large | O60A812E | Each |
| 17150975 | Medium | O60A819E | Each |
| 17160915 | Large | O60T802E | Each |
| 17160935 | 2X-Large | O60T808E | Each |
| 17160955 | Small | O60A813E | Each |
| 17160975 | Large | O60A819E | Each |
| 17170915 | X-Large | O60T802E | Each |
| 17170935 | 3X-Large | O60T808E | Each |
| 17170955 | Medium | O60A813E | Each |
| 17170975 | X-Large | O60A819E | Each |
| 17180905 | Small | O60T801B | Each |
| 17180915 | 2X-Large | O60T802E | Each |
| 17180935 | Small | O60T809E | Each |
| 17180955 | Large | O60A813E | Each |
| 17180965 | Small | O60A818E | Each |
| 17180975 | 2X-Large | O60A819E | Each |
| 17190905 | Medium | O60T801B | Each |
| 17190935 | Medium | O60T809E | Each |
| 17190955 | X-Large | O60A813E | Each |
| 17190965 | Medium | O60A818E | Each |
| 17190975 | 3X-Large | O60A819E | Each |
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Descrizione
- Excellent chemical protection, >480 mins for 15 chemicals specified by EN 943-2:2002
- Innovative fabric technology, high performance barrier laminate with scrim reinforcement
- Lightweight and flexible
- Highly visible bright orange color for improved worker safety
- Ultrasonically welded and hot air taped seams
- DYNAT/YKK Gas-Tight zipper provides protection and performance in the most hostile of chemical environments
- The zip has an outer zip flap made of the same material as the suit
- Semi-rigid 3 layer GAG visor provides excellent optical clarity
Specifica
Specifica
| Color | Orange |
| For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases, Tank Cleaning |
| Size | 3X-Large |
| Style | O60T802E |
| Weight (Metric) | 230 gsm |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.