missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Micrococcal Nuclease (300 U/μL)

Product Code. 10848990 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
8,000 units
Unit Size:
8000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10848990 8,000 units 8000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10848990 Supplier Thermo Scientific™ Supplier No. EN0181
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Digest single-stranded and double-stranded DNA and RNA with Thermo Scientific™ Micrococcal Nuclease (S7 Nuclease), an endo-exonuclease.

Micrococcal Nuclease (S7 Nuclease) is a relatively nonspecific endo-exonuclease that digests single-stranded and double-stranded nucleic acids, but is more active on single-stranded substrates. Cleavage of DNA or RNA occurs preferentially at AT or AU-rich regions, yielding mononucleotides and oligonucleotides with terminal 3'-phosphates. The enzyme's activity is strictly dependent on Ca2+.

Applications

  • Hydrolysis of nucleic acids in crude cell-free extracts
  • Sequencing of RNA
  • Studies of chromatin structure
  • A model for protein folding and for structure-function studies
TRUSTED_SUSTAINABILITY

Specifications

Concentration 300 U/μL
Enzyme Nuclease
Quantity 8,000 units
Product Type Micrococcal Nuclease

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.