missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Microflex™ Ergonomic Neoprene Disposable Gloves

Product Code. 15589451
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15589451 Small Pack of 100
15509461 Large Pack of 100
15519461 X-Large Pack of 100
15579451 X-Small Pack of 100
15599451 Medium Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15589451 Supplier Ansell Edmont™ Supplier No. 73847070
  • ONLINE EXCLUSIVE PRICE
    34.86 €
    22.09 € / Pack of 100
    Save 12.77 €
    36% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Soft and pliable neoprene formulation provides a more comfortable wear for longer periods, while allowing workers greater tactile sensitivity. Microflex 73-847 gloves are proven to reduce muscle effort and hand fatigue while enhancing gripping abilities in wet conditions.

  • Provide a more comfortable wear for longer periods
  • Textured fingers ensure a strong and secure grip
  • Deliver excellent wet and dry grip
  • Made without natural rubber proteins and powder to help protect workers from allergies, skin irritation and dryness
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 13485, PPED 89/686/EEC CATEGORY III, COMPLEX DESIGN, EN374-3, EN374-2 LEVEL 2 AQL 1.5, ASTM D6977, EN420
Applications Laboratory
Color Green
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Neoprene
Powdered Powder Free
Product Type Neoprene Gloves
Size Small
Sterility Non-sterile
Thickness (Metric) 0.10 mm
Finger Style Textured
Glove Cuff Style Beaded/Rolled
Model 73-847
Packaging Type Dispenser Box
Quantity 100 Pk.
Tensile Strength 18 MPa
Length (Metric) 245 mm
Width (Metric) 86 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.