missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes
Shop All Agilent BioTek Products

Click to view available options
Diameter (Metric):
0.02 cm
0.035 cm
Type:
Large Volume Replacement Tubing Set
Low Volume Replacement Tubing Set
Medium Volume Replacement Tubing Set
Material:
Molded Polypropylene
Capacity (Metric):
1 to 50 μL
10 to 3000 μL
5 to 2500 μL
missing translation for 'unitSize'
Each
Description
- Volume range 1 μL to 10 mL
- Accommodates 6- to 1536-well microplates, PCR trays, deep-well microplates, microtubes and other tube configurations up to 4 in. in height
- Dispense 1536 wells in 18 s.
- Cassettes can be autoclaved without recalibration
- Dispense up to eight reagents at one time
- Customizable dispense pattern allows skipped columns or rows
- Built-in priming trough
- Easy maintenance of individually replaceable tips and choice of replacement cassettes or tubing
- Compatible with Bio-Stack microplate stacker 11-120-601 for increased throughput
Spécification
Spécification
| Capacity (Metric) | 1 to 50 μL |
| Diameter (English) | 0.01 in. |
| Diameter (Metric) | 0.02 cm |
| For Use With (Application) | For liquid handling |
| Increments | 1 μL |
| Material | Molded Polypropylene |
| Type | Low Volume Replacement Tubing Set |
| For Use With (Equipment) | EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu