Learn More
Ansell Microgard™ 1800 Standard Coveralls with Hood
Product Code. 15546504
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 40
Pack of 50
Product Code. 15546504
Brand: Ansell WH18B0011105
Only null left
Add to basket
Add to basket
Provides comfort in warmer working environments. Ansell Microgard™ 1800 Standard Coveralls with Hood are constructed of lightweight, breathable material yet protects wearer from light spray and dry particles.
  • Coverall with 3-piece hood
  • Elasticated wrists, waist and ankles
  • 2-way front zipper with re-sealable storm flap
  • Bound seams
  • Standard: CE category III, type 5/6 EN 1149-5 (antistatic)
  • Low hazard liquid spray, splash and particle protection
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Silicone-free which is critical in spray painting applications
  • Anti-static according to EN1149-5
  • Low linting to reduce the risk of fibre contamination in some critical areas
  • Optimised body fit for wearer comfort and safety
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size X-Large
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance (Europe) CE category III, type 5/6 EN 1149-5
Model WH18-B-00-111
No. of Pockets None
Style Includes Hood