Learn More
Ansell Microgard™ 2000 Standard White Coveralls with Hood
Product Code. 15556514 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 40
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15556514 Large Pack of 40
15536514 Small Pack of 40
15546514 Medium Pack of 40
15566514 X-Large Pack of 40
15576514 2X-Large Pack of 40
15586514 3X-Large Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15556514 Supplier Ansell Supplier No. WH20B0011104
  • ONLINE EXCLUSIVE PRICE
    459.90 €
    300.88 € / Pack of 40
    Save 159.02 €
    34% Off
View all promotions
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Allows perspiration to escape from the suit yet withstands saturation of liquid chemicals. Ansell Microgard™ 2000 Standard White Coveralls with Hood filter 100% of particulates down to 0.01 microns in size.
  • Coverall with 3-piece hood
  • White bound seams
  • Elasticated hood, waist, wrists and ankles
  • Includes fingerloops
  • 2-way front zipper with re-sealable storm flap.
  • Liquid penetration resistance and barrier to fine particulates (>0.01 microns)
  • High quality two-way stretch microporous film provides an effective liquid and particle barrier
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Silicone free - Critical in paint spraying applications
  • Low linting - Reduces the risk of fibre contamination in some critical areas
  • Optimized body fit provides wearer comfort and safety
  • Anti-static treatment
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size Large
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Bound
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance (Europe) Cat III Type 5-B, 6-B, EN1073-2, EN14126, EN1149-1, DIN 32781
Model WH20-B-00-111
No. of Pockets None
Style Includes Hood, Fingerloops