Learn More
Ansell Microgard™ 2000 Standard White Coveralls with Hood and Boots
Product Code. 15516524 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 30
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15516524 Large Pack of 30
15506524 Medium Pack of 30
15596514 Small Pack of 30
15526524 X-Large Pack of 30
15536524 2X-Large Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15516524 Supplier Ansell Supplier No. WH20B0012204
  • ONLINE EXCLUSIVE PRICE
    443.10 €
    290.38 € / Pack of 30
    Save 152.72 €
    34% Off
View all promotions
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to allow water vapor (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals. Filter 100% of particulates down to 0.01 microns in size.
  • Coverall with 3-piece hood and boots with anti-slip soles
  • White bound seams
  • Elasticated hood, waist and wrists
  • Includes finger loops
  • 2-way front zipper with resealable storm flap
  • Anti-static treatment
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance (Europe) Cat III Type 5-B, 6-B, EN1073-2, EN14126, EN1149-1, DIN 32781
Description Hood, boots, and finger loops
Model WH20-B-00-122
No. of Pockets None
Style Includes Hood, Boots, and Fingerloops