missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Microgard™ 2000 Ts Plus Coveralls with Hood
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 45
Description
- Proven barrier to low concentration liquid chemicals, diluted pesticides, liquid and particulate biological hazards
- Moisture vapor permeable to help reduce the risk of heat stress
- Silicone-free for spray painting applications
- Low Linting to reduce the risk of contamination in critical areas
- Anti-static according to EN 1149-1
- Optimized body fit for wearer comfort and safety
- Tunnelled elasticated wrists, hood and ankles minimises the risk of linting and cross contamination
- Thumb loops prevent sleeve movement when working above overhead
- Chinstrap helps to reduce the risk of cross contamination
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | Medium |
| Gender | Unisex |
| Material | Microporous Polyethylene Film |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction