Learn More
Ansell Microgard™ 2500 Standard Overshoes
Description
- Elasticated opening
- Bound seams
- Microporous polypropylene laminate
- Fits Euro sizes 42 to 46 (UK 8 to 11)
Specifications
Specifications
| Material | Polypropylene |
| Color | White |
| Closure Type | Elastic Opening |
| Model | WH25-B-00-400-00 |
| Seam Type | Bound |
| Packaging Quantity | 350 Pairs |
| Product Type | Shoe Covers |