Learn More
Ansell™ Micrograd™ MICROCHEM™ 5000 Coveralls with Hood
Product Code. 15556474
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 10
Product Code. 15556474
Brand: Ansell™ OR50T0011104
Only null left
Add to basket
Add to basket
Micrograd™ MICROCHEM™ 5000 Coveralls with Hood reaches new levels in chemical protection and has been engineered to protect. The highly visible multi-layer fabric is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams
  • Coverall with 2-piece hood
  • Elasticated double cuffs (knitted inner cuff), waist and ankles
  • Seams are ultrasonically welded and taped
  • Highly visible bright orange colour for improved worker safety
  • Liquid-tight dual zip designs without the need for additional taping
  • Provides a barrier to numerous organic and inorganic chemicals and biological hazards
  • Comfortable multi-layer material is lightweight, yet strong and durable
  • Protection - >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size Large
Gender Unisex
Material Multi-layer Fabric
Color Orange
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Knit
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Liquid-tight Double Zipper
Seam Type Taped, Welded
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance Cat. III Type 3-B, 4-B, 5-B, EN1073-2, EN14126, EN1149-1
Description Elasticated with double knitted inner cuff
No. of Pockets None
Style Includes Hood