missing translation for 'onlineSavingsMsg'
Learn More

BD Microlance™ Stainless Steel Needles

Color coded for easy recognition of the needle’s guage. BD™ Microlance™ Stainless Steel Needles feature thin walls for optimal flow and fit Luer slip syringes. These sterile, individually wrapped needles ensure more comfortable insertion with less risk for punching.

Brand:  BD 300900

Code : NEW

Additional Details : Weight : 0.10710kg

 View more versions of this product

Product Code. 10020464

  • 10.86 € / Pack of 100
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Description

Description

Unique slip angle and polishing method results in less patient discomfort at the injection site and less risk for punching.

  • Hypodermic needles with thin walls for optimal flow
  • Fits syringes connected to a Luer adapter or Luer Lock connector
  • Color coded for quick recognition of gauge for easy selection of appropriate flow rate
  • Sterile and individually wrapped for hygienic, single-use application
  • Stainless steel needles with epoxy resin adhesives
Specifications

Specifications

Black
22 G
For single use only
1.25 in.
Bevel
Sterile
5,000 individually wrapped needles, thin cannula wall
0.7 mm
Yes
Needle
Connected to a Luer adapter or Luer Lock connector
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers