missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Isolator Tool and Irradiated Covers

The irradiated angled head adapter with a 360° swivel works with a range of irradiated mop covers to efficiently clean and disinfect isolators, hoods, RABS and small equipment in clean and aseptic environments.

Brand:  Micronova™ ITC620-911IR

Code : 04

Additional Details : Weight : 1.70000kg

 View more versions of this product

Product Code. 13118675

  • 665.00 € / Pack of 50
Estimated Shipment: 27-09-2024
to see stock.
Add to basket

Description

Description

Help to simplify cleaning in compliance with USP 797

The Micronova Irradiated Isolator Tool is designed for use with the SSU Series Universal Handles or STA Series threaded handles to fit through small passes and allow extended reach. The mop covers are available in a range of fabrics with optional foam layer to hold disinfectant and facilitate wringing.

  • Irradiated plastic isolator tool with choice of irradiated covers
  • Ergonomic cleaning for isolators, hoods, RABS and other small equipment
  • Modular and extendable handles fit through small passes and expand reach
  • 360° swivel, angled head adapter for easy application of cleaning agent
  • Gamma irradiated for use in aseptic environments
  • Covers available in a variety of fabrics for specialized cleaning
    • NovaPoly — 100% smooth knit polyester provides excellent disinfectant dispersal
    • PolySorb — 100% textured polyester for removing residues and dispersing disinfectant
    • MegaTex — 100% non-woven polyamide is absorbent with a mildly abrasive surface
  • Compatible with alcohol, strong disinfectants and steam sterilization
  • Launderable for reuse
  • Autoclavable
Specifications

Specifications

22.9 x 5.1 cm (9 x 2 in.)
White
Sterile
Autoclavable
2 in.
Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
Non-woven Polyamide
Isolator Cleaning Tool Cover
9 in.
22.9 cm
5.1 cm
No
For Use with Isolators, Containment, and Bio-Hoods in Cleanroom and Buffer Areas
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers