Learn More
Thermo Scientific™ Microplate Reader Verification Plates
Shop All Thermo Scientific Products
Click to view available options
Product Type:
Multiskan Verification Plate, 16 wavelengths
Multiskan Verification Plate, 8 wavelengths
Spectrophotometric Verification Plate
Verification Plate
Unit Size:
Each
Description
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Multiskan Verification Plates and Spectrophotometric Verification Plate enables you to verify parameters such as photometric accuracy, precision and linearity, as well as wavelength accuracy and stray light radiation.
Lumiwell Luminometric Verification Plate is designed to be an independent Relative Light Unit reference for luminometry with the Thermo Scientific luminescence microplate readers.
We recommend recalibrating the verification plates regularly. The frequency of calibration is dependent on use and laboratory procedures. However, the recalibration should be performed at least every two years.
Lumiwell Luminometric Verification Plate is designed to be an independent Relative Light Unit reference for luminometry with the Thermo Scientific luminescence microplate readers.
We recommend recalibrating the verification plates regularly. The frequency of calibration is dependent on use and laboratory procedures. However, the recalibration should be performed at least every two years.
Specifications
Specifications
| Product Type | Verification Plate |
| For Use With (Equipment) | Varioskan LUX Microplate Reader, Luminoskan |
| Plate Type | Verification Plate |
| Includes | Lumiwell Luminometric Verification Plate Certificate of Calibration Carrying case Lumiwell Verification Plate |