Learn More
Thermo Scientific™ Microplate Reader Verification Plates
Product Code. 17104669 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Multiskan Verification Plate, 16 wavelengths
Multiskan Verification Plate, 8 wavelengths
Spectrophotometric Verification Plate
Verification Plate
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
17104669 Multiskan Verification Plate, 16 wavelengths Each
11958995 Multiskan Verification Plate, 8 wavelengths Each
12463458 Verification Plate Each
11560715 Spectrophotometric Verification Plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17104669 Supplier Thermo Scientific™ Supplier No. 24072805
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Thermo Scientific Microplate Reader Verification Plates are dedicated performance verification systems helping to verify and document the key performance parameters of Thermo Scientific Multiskan FC, Multiskan Sky, Luminoskan, Fluoroskan FL, and Varioskan LUX.
Multiskan Verification Plates and Spectrophotometric Verification Plate enables you to verify parameters such as photometric accuracy, precision and linearity, as well as wavelength accuracy and stray light radiation. 
Lumiwell Luminometric Verification Plate is designed to be an independent Relative Light Unit reference for luminometry with the Thermo Scientific luminescence microplate readers.
We recommend recalibrating the verification plates regularly. The frequency of calibration is dependent on use and laboratory procedures. However, the recalibration should be performed at least every two years.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multiskan Verification Plate, 16 wavelengths
For Use With (Equipment) Multiskan FC
Plate Type Verification Plate
Product Line Multiskan
Includes NPL (National Physical Laboratory) traceable Multiskan™ Verification Plate
Certificate of Calibration
USB memory device including SkanIt™ software sessions and reference files
Carrying case for Multiskan™ Verification Plate