missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Microplate Rotor for Thermo Scientific Sorvall™ Legend™ Mach 1.6™/1.6R Centrifuges

For use with Legend Mach 1.6 and 1.6R Centrifuges.

Brand:  Thermo Scientific™ 75002010

Code : 51

Additional Details : Weight : 2.50000kg

Product Code. 10533633

  • 3555.00 € / Each
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Rotor and lid

Description

Description

  • Capacity: Four microplates or two deepwell plates
  • Max. Speed/RCF: 4400rpm/3355 × g
  • Biocontainment-approved with optional lid 75-002-011 (Sold separately, in pairs)
  • Microplates sold separately

Specifications

Specifications

MP 3300 Microplate Rotor
3,355 x g
4,400 rpm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions