missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Microsart™ Bacterial DNA Extraction Kit
Shop All Sartorius Products
Click to view available options
:
Reagents for 50 extraction
Description
- Microsart™ Bacteria Extraction kit introduces a state-of-the-art DNA extraction method for DNA extraction from cell cultures and cell culture derived biologicals, like autologous transplants and other advanced therapy medicinal products (ATMP), for subsequent DNA amplification via PCRh.
- Eliminates the risk of DNA contaminations, facilitating the detection of bacteria in cell culture and ATMPs via PCR.
Specifications
Specifications
| For Use With (Application) | DNA Extraction |
| No. of Reactions | Reagents for 50 extraction |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu