Learn More
EDU Lab™ Microscience™ Microburette Kit
Product Code. 12806955
Change view
Click to view available options
For Use With: Use in conjunction with MicroChem kit Microscience
Product Code.
12806955 For Use With: Use in conjunction with MicroChem kit Microscience
1 options
Product Code. 12806955 Supplier EDU Lab™ Supplier No. 310012
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
For Use With (Application) Use in conjunction with MicroChem kit Microscience
Includes Standardization of hydrochloric acid using sodium carbonate; standardization of hydroxide using standard hydrochloric acid; calcium and magnesium ion concentration; a conductometric titration
Product Type MicroChem Microburette Kit