Description
- Supplied complete with two sliding filter holders, one containing the blue and green filter blocks, the other the violet and UV filter blocks
- Objectives consist of 10x, 25x and 40x for both brightfield and fluorescent viewing, plus 10x, 25x and 40x for phase contrast with the corresponding annular rings
- Quintuple revolving nosepiece with click stop
- Includes spare fuse, green/blue transmitted light filters, microtiter stage frame, rubber eye cups, dust cover and instruction manual
Specifications
Specifications
| Includes | Spare fuse, green and blue transmitted light filters, microtitre stage frame, rubber eye cups, dust cover and instruction manual |
| Nosepiece | Quintuple revolving with click stop, inverted |
| Weight (Metric) | 24 kg |
| Lighting Type | Reflected Light 100w HBO External Source with Controller |
Product Title
¿Detecta una oportunidad de mejora?