Learn More
DuraLab Microscope Objective 100x for DF-50
Description
- Provides high magnification for observing cells and microorganisms in detail
- Spring-loaded lens ensures safe handling with immersion oil
- Supports advanced microscopy skills and detailed biological studies
- Aligned with UK science curriculum for in-depth biological exploration and preparation for higher education
- Compatible with 20x eyepiece to further increase total magnification
Specifications
Specifications
| Type | Microscope Objective |
| For Use With (Equipment) | DRL0038 |
Compatible with DRL0038 - DuraLab DF-50 Classic Monocular Microscope