Learn More
Heinz Herenz Microscope Slides with Cut Edges
Product Code. 15502304
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 100
Product Code. Color Unit Size
15502304 Black Pack of 100
1 options
Product Code. 15502304 Supplier Heinz Herenz Supplier No. 1041015
  • ONLINE EXCLUSIVE PRICE
    106.05 €
    74.00 € / Pack of 100
    Save 32.05 €
    30% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Microscope slides, economy, with cut edges. Transparent green glass.
We offer a very wide range of slides, so you are sure to find what you are looking for.
  • Within the product lines you will find cut and ground slides.
  • Cut slides are preferred for routine microscopy and offer great flexibility.
  • Ground slides prevent cuts due to the ground edges.
  • If you want to label and categorize your slides, it is best to choose slides with matted edges or a coloured field.
  • You also have the choice between slides with and without cellophane.
Specifications
Width (Metric) 8 mm
Sterility Non-sterile
Edges Ground
Material PTFE
Type Diagnostic Slide
Well Count 10
Color Black