Learn More
Applied Biosystems™ MicroSEQ™ ID 16S rDNA 500 Library v2022
Product Code. 18723634 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity unitSize
18723634 1 Ea. Each
1 options
Product Code. 18723634 Supplier Applied Biosystems™ Supplier No. A57312
The MicroSEQ ID 16S rDNA 500 Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0.
The MicroSEQ ID 16S rDNA 500 Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0. It contains over 2100 reference 16S rRNA sequences from bacteria. The library entries contain the first 500 bases of the 16S rRNA gene, enough to identify bacterial unknowns.
TRUSTED_SUSTAINABILITY
Specifications
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers
For Use With (Application) Microbial Identification
License Static
Operating System Microsoft Windows 10
Product Line MicroSeq™ ID
Product Type Microbial Identification Software
Quantity 1 Ea.
Software Data Collection and Analysis Software