Learn More
Applied Biosystems™ MicroSEQ™ ID 16s rDNA 500 Supplemental Library v2022
Product Code. 18783634 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity unitSize
18783634 1 Ea. Each
1 options
Product Code. 18783634 Supplier Applied Biosystems™ Supplier No. A57313
The MicroSEQ ID 16S rDNA 500 Supplemental Library v2022 is a stand-alone library that extends the size of the MicroSEQ ID 16S rDNA 500 Library to strengthen the identification process when using MicroSEQ ID Microbial Identification Software.
The MicroSEQ ID 16S rDNA 500 Supplemental Library v2022 is a stand-alone library that extends the size of the MicroSEQ ID 16S rDNA 500 Library to strengthen the identification process when using MicroSEQ ID Microbial Identification Software. The Supplemental Library contains 7,138 16S rDNA public sequences of type and non-type strains of bacterial species that underwent the same rigorous validation process as the sequences in the MicroSEQ ID 16S rDNA 500 Library. The Supplemental Library is used when the MicroSEQ ID 16S rDNA 500 Library results in a percent match lower than the species ID acceptance criteria.
Key features include:
Quality controlled—an in-silico validated library with over 7,000 type and non-type strains quality-checked for sequence, taxonomy, nomenclature, and phylogeny
Simplified—reliable entries from public resources converted into a simple format for routine validation of unique environmental isolates
Consistent—the same acceptance criteria for analysis and the same reporting format in the software
Maintained—regular updates for entries and information supported by experienced Thermo Fisher Scientific technical staff
TRUSTED_SUSTAINABILITY
Specifications
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers
For Use With (Application) Microbial Identification
License Static
Operating System Microsoft Windows 10
Product Line MicroSeq™ ID
Product Type Microbial Identification Software
Quantity 1 Ea.
Software Data Collection and Analysis Software