Learn More
Applied Biosystems™ MicroSEQ™ ID 16S rDNA Full Gene Library v2.1
Shop All Applied Biosystems ProductsDescription
The MicroSEQ ID 16S rDNA Full Gene Library v2.1 is designed for installation with the MicroSEQ ID analysis software. It contains the complete 16S sequence from bacterial organisms that can be compared to an unknown bacterial sequence using MicroSEQ ID analysis software to identify bacteria. MicroSEQ ID analysis software enables you to identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library.
• A validated library of over 2300 microbial sequences ensures you get the answers you need
• Easy-to-use interface and optimized search algorithms enable quick results
• Phylogenetic trees provide a deeper understanding of your unknowns
• Security features support 21 CFR Part 11 compliance, enabling the software to integrate smoothly into your workflow
Specifications
Specifications
| Sequencing Type | Genome and DNA Sequencing |
| For Use With (Equipment) | SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers |
| For Use With (Application) | Microbial Identification |
| License | Static |
| Operating System | Microsoft Windows 10 |
| Product Line | MicroSeq™ ID |
| Product Type | Microbial Identification Software |
| Quantity | 1 Ea. |
| Software | Sequence Analysis, Microbial Identification |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.