Learn More
MicroSEQ™ ID Fungal Gene Library, v2018
Product Code. 18886655 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. Quantity Unit Size
18886655 1 each Each
1 options
Product Code. 18886655 Supplier ABI Applied Biosystems Supplier No. A47135
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The MicroSEQ ID Fungal Gene Library v2018 is for use with MicroSEQ ID Microbial Identification Software v3.0 and v3.1.
The MicroSEQ ID Fungal Gene Library v2018 is for use with MicroSEQ ID Microbial Identification Software v3.0 and v3.1. It contains over 1300 reference sequences from the D2 region of the Large Subunit (rDNA) of yeast and mold species against which an unknown yeast or mold sequence can be compared in order to identify the unknown organism. This library is delivered in either digital or USB format.
TRUSTED_SUSTAINABILITY
Specifications
Product Line MicroSEQ™ ID
Quantity 1 each
Shipping Condition Room Temperature